missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epT.I.P.S.™ 384 Reloads - Micropipette Tips

Product Code. 16212381 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 to 20 μL
5 to 100 μL
Clean Claims:
Eppendorf Quality
PCR Clean
Unit Size:
Pack of 3840
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Clean Claims Unit Size
16212381 0.1 to 20 μL Eppendorf Quality Pack of 3840
16272371 0.1 to 20 μL PCR Clean Pack of 3840
16282371 5 to 100 μL PCR Clean Pack of 3840
16222381 5 to 100 μL Eppendorf Quality Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16212381 Supplier Eppendorf™ Supplier No. 0030076044
  • ONLINE EXCLUSIVE PRICE
    468.33 €
    418.00 € / Pack of 3840
    Save 50.33 €
    10% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eppendorf epT.I.P.S.™ 384 Reloads provide high-precision micropipette tips in 'Eppendorf Quality' and 'PCR clean' grades, tailored for 384-well plate applications, ensuring accuracy, reduced waste, and compatibility in high-throughput labs.

  • epT.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16- and 24-channel pipettes as well as 20 μL and 100 μL Move It adjustable tip spacing pipettes for utmost safety and reliability
  • Designed for a cone distance of 4.5 mm
  • Innovative SOFTattach technology allows for reliable tip fit and tightness
  • Fill a whole 384-well plate without loosening pipette tips
  • Perfect tip alignment by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of the epT.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Tip attachment forces reduced by 40% per cone compared to 8- and 12-channel pipettes
  • Tip ejection forces in 16- and 24-channel pipettes reduced by SOFTeject for ergonomic working conditions
  • Available as reload, and set
  • Available in purity grades of Eppendorf Quality and PCR clean
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Model epT.I.P.S.™ 384 Reloads
Length (English) 1.7 in.
Length (Metric) 42 mm
Clean Claims Eppendorf Quality
Storage Requirements Room temperature
Format Refill
Tip Style Universal
Packaging 3,840 Tips (10 Trays x 384 Tips)
Quantity 3840 Tips
Product Type Micropipette Tips
Color Code Light Pink
Disposable Single-use
For Use With (Equipment) Air-Cushion System, Universal
Product Line epT.I.P.S.
Material Polypropylene
Show More Show Less

The pipette tips of the :Productname: product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned :Productname: are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.