missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf epT.I.P.S.™ Motion as SafeRack tips

Product Code. 30019026 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
1.000 μL
50 μL
300 μL
Sterility:
Non-sterile
PCR Clean
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Unit Size
30019026 1.000 μL PCR Clean Filtered Pack of 960
30019023 50 μL Non-sterile Non-filtered Pack of 960
30019050 50 μL PCR Clean Filtered Pack of 960
30019025 300 μL Non-sterile Non-filtered Pack of 960
30019055 300 μL PCR Clean Filtered Pack of 960
30019047 1.000 μL Non-sterile Non-filtered Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30019026 Supplier Eppendorf Supplier No. 0030014650
  • ONLINE EXCLUSIVE PRICE
    263.00 €
    244.00 € / Pack of 960
    Save 19.00 €
    7% Off
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well.

  • Specially designed for use on the epMotion™ 5070 and 5075 Liquid Handling Workstation. epT.I.P.S. Motion SafeRack pipette tips, with and without filters
  • Developed in torsion-resistant, contamination-free racks that are recommended for the re-use of tips feature of epMotion 5073 systems.

Reuse Tips Feature:

  • The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well
  • Pipette tips will be placed back into the position of the box they were removed from
  • Returned tips will be reused within an application for the same source vessel
  • Individual compartments in rack to avoid cross-contamination between used tips
  • Available as standard and PCR clean quality for 50, 300, and 1000 μL
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1.000 μL
Filtered Filtered
Sterility PCR Clean
Color Clear
Autoclavable Not Autoclavable
Format Box
Tip Style Standard, Straight
Product Type Robotic Tips
Disposable Single-use
For Use With (Equipment) Eppendorf epMotion
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.