Learn More
Description
- epT.I.P.S.™ Box 2.0 (EU-IVD), Eppendorf Quality™
- Contamination-free transfer of trays to stable working box
- System optimized for use with multichannel pipettes
- Color-coded trays for simple identification of tips and pipette
- Tips can be attached to pipette from refill trays
- The trays and boxes can be autoclaved at 121 °C for subsequent use
- The pipette tips of the epT.I.P.S.™ product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned epT.I.P.S.™ are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.
Specifications
Specifications
| Volume (Metric) | 50 to 1000 μL |
| Filtered | Non-filtered |
| Color | Colorless |
| Autoclavable | Autoclavable |
| Length (Metric) | 71 mm |
| Certifications/Compliance | 98/79 EC |
| Clean Claims | Eppendorf Quality™ |
| Packaging | Set |
| Quantity | 480 Pk. |
| Product Type | Consumables |
| Show More |
The pipette tips of the epT.I.P.S.™ product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned epT.I.P.S.™ are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
