Learn More
Eppendorf™ GELoader™ Tips
GREENER_CHOICE
Product Code. 10411193
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
Unit Size:
Pack of 192
Product Code. Volume (Metric) unitSize
10411193 0.5 to 20 μL Pack of 192
1 options
Product Code. 10411193 Supplier Eppendorf™ Supplier No. 0030001222
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these flexible long and narrow pipette tips for handling of smallest volumes. Eppendorf GELoader™ Tips are ideal for gel electrophoresis and loading of polyacrylamide gels.
  • GELoader tips and rack autoclavable
  • Highest precision and accuracy when used with Eppendorf pipettes for 0.5 to 10μL (gray control button)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 20 μL
Autoclavable Autoclavable
Length (Metric) 6.8cm
For Use With (Application) For loading of samples onto polyacrylamide gels
Tip Style Gel Loading
Product Line GELoader
Material Polypropylene