missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Equine Donor Serum

Product Code. 11485035 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Color:
Golden Brown
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Color Unit Size
11485035 100 mL Golden Brown 100mL
11495035 500 mL Golden Brown 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485035 Supplier MP Biomedicals™ Supplier No. 092921149
  • ONLINE EXCLUSIVE PRICE
    33.25 €
    31.50 € / 100mL
    Save 1.75 €
    5% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance U.S. regulations
Color Golden Brown
Content And Storage -20°C
Species Horse
Form Liquid
Product Type Horse Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method 0.2 μm filtration
For Use With (Application) Diagnostic assays
Hemoglobin Concentration ≤35mg/dL
Mycoplasma Detected No
Osmolality 273 to 301mOsm/kg
pH 7.1 to 8
Protein Content 5.6 to 9.1 g/dL
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.