missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PolarScreen™ ER Alpha Competitor Assay, Red

Product Code. 15607858
Change view
Click to view available options
Quantity:
800 assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15607858 800 assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15607858 Supplier Thermo Scientific™ Supplier No. A15884
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PolarScreen™ ER Alpha Competitor Assay, Red provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands.

  • The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands. The kit uses insect cell-expressed, full-length, native (untagged) human estrogen receptor alpha (available separately, Cat. No. A15674) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone EL Red) in a homogenous, mix-and-read assay format.
  • This kit produces a red color read-out; a green kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation.
  • Complete & ready-to-use out of the box - all reagents included, just add your test compounds
  • High throughput compatible - 800 x 20 μL fluorescence polarization assays in a 384-well plate
  • Convenient - available in bulk sizes for larger screening needs
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:

• 1 tube full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
• 1 tube of Fluormone EL Red Tracer (store at -5 to -30°C)
• 2 bottles of ER Red Screening Buffer (store at -5 to -30°C)
Detection Method Fluorescence
For Use With (Equipment) Fluorescent Imager
Format 384-well Plate
Quantity 800 assays
Type ER Alpha Competitor Assay

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.