missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ERB 041
GREENER_CHOICE

Product Code. p-4816695
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18055412

Brand: Tocris Bioscience™ 4276/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent ERβ agonist

Potent ERβ agonist; displays >200-fold selectivity for ERβ over ERα (IC50 values are 5 and 1216 nM for human ERβ and ERα; 3.1 and 620 nM for rat ERβ and ERα; and 3.7 and 750 nM for mouse ERβ and ERα respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524684-52-4
Molecular Formula C15H10FNO3

Specifications

Quantity 10 mg
Formula Weight 271.24
Purity >99%
Molecular Weight (g/mol) 271.247
Chemical Name or Material ERB 041
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.