missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Product Code. 11356668
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356668 2 g 2g
11366668 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356668 Supplier Thermo Scientific Alfa Aesar Supplier No. 013214.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14553-08-3 | C15H21ErO6 | 464.59 g/mol

Erbium(III) 2,4-pentanedionate is commonly used in various catalysts and catalytic reagents for organic synthesis. It is also used for glass coloring, as an amplifier in fiber optics, and in lasers for medical and dental purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14553-08-3
Molecular Formula C15H21ErO6
Molecular Weight (g/mol) 464.59
MDL Number MFCD00013490
InChI Key FGQSJRDKBCVFHH-UHFFFAOYSA-N
Synonym erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one
PubChem CID 102601666
SMILES [Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 2 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
IUPAC Name erbium(3+) tris(2,4-dioxopentan-3-ide)
Formula Weight 464.59
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Erbium(III) 2, 4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.