missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) isopropoxide

Product Code. 11318058
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11318058

Brand: Thermo Scientific Alfa Aesar 036613.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14814-07-4 | C9H21ErO3 | 344.523 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14814-07-4
Molecular Formula C9H21ErO3
Molecular Weight (g/mol) 344.523
MDL Number MFCD00144253
InChI Key VOCNVTAXVORJBI-UHFFFAOYSA-N
Synonym erbium iii isopropoxide, triisopropoxyerbium, erbium-isopropoxide, acmc-20al30, erbium iii i-propoxide, erbium 3+ ; propan-2-olate, erbium 3+ ion tris propan-2-olate, erbium 3+ tris propan-2-olate
PubChem CID 2734042
IUPAC Name erbium(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Er+3]

Specifications

Melting Point 325°C (decomposition)
Flash Point 58°C (138°F)
Odor Alcohol-like
Linear Formula Er[OCH(CH3)2]3
Quantity 1 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Formula Weight 344.52
Physical Form Powder
Chemical Name or Material Erbium(III) isopropoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.