missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxalate decahydrate, 99.9%

Product Code. 11438347
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11438347

Brand: Thermo Scientific Alfa Aesar A18420.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30618-31-6 | C6H26Er2O22 | 784.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30618-31-6
Molecular Formula C6H26Er2O22
Molecular Weight (g/mol) 784.77
MDL Number MFCD00150142,MFCD00150142
InChI Key CYHKKMLSGVLNPD-UHFFFAOYSA-N
Synonym erbium oxalate decahydrate,
PubChem CID 131877482
IUPAC Name erbium;oxalic acid;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O

Specifications

Density 2.64
Linear Formula Er2(C2O4)3·10H2O
Quantity 100 g
UN Number UN3288
Sensitivity Hygroscopic
Formula Weight 778.73 (598.58 Anhydrous)
Percent Purity 99.9%
Chemical Name or Material Erbium(III) oxalate decahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.