missing translation for 'onlineSavingsMsg'
Learn More

Erbium pieces, 99.9% (REO)

Product Code. p-4781895
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle under Argon
Packaged under argon in glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11370659

Brand: Thermo Scientific Alfa Aesar 000111.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-52-0 | Er | 167.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Specifications

Color Silver to Gray
Appearance Silver Gray pieces
Density 9.066 g/cm3
Quantity 10 g
Packaging Glass Bottle under Argon
Assay Percent Range 99.9% (REO)
Form Pieces
Chemical Name or Material Erbium, Rare Earth Oxides Content

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.