missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ergosterol, 98%

Product Code. 10500591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10500591 5 g Glass bottle 5g
10677642 25 g Glass bottle 25g
10381044 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10500591 Supplier Thermo Scientific Chemicals Supplier No. 117810050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-87-4 | C28H44O | 396.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-87-4
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.64
MDL Number MFCD00003623
InChI Key DNVPQKQSNYMLRS-APGDWVJJSA-N
Synonym ergosterol, provitamin d2, unii-z30ray509f, ergosterin, ergosta-5,7,22e-trien-3beta-ol, 5,7,22-ergostatrien-3beta-ol, 3beta-hydroxy-5,7,22-ergostatriene, 22e,24s-24-methylcholesta-5,7,22-trien-3beta-ol, 22e-ergosta-5,7,22-trien-3beta-ol, ergosta-5,7,22-trien-3-ol, 3b,22e
PubChem CID 444679
ChEBI CHEBI:16933
IUPAC Name (3S,9S,10R,13R,14R,17R)-17-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)C(C)C=CC(C)C1CCC2C1(CCC3C2=CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 156°C to 158°C
Color White
Boiling Point 250°C (1.3 Pa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 06,III,3099
Merck Index 15, 3716
Specific Rotation Condition −120.00 (20.00°C c=1,CHC13)
Specific Rotation -120
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in benzene,1g/660 mL alcohol- 1g/45 mL boiling alcohol,1g/70 mL ether- 1g/39 mL boiling ether,1g/31 mL chloroform
Formula Weight 396.64
Percent Purity 98%
Physical Form Crystalline Powder or Crystalline Needles
Chemical Name or Material Ergosterol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.