missing translation for 'onlineSavingsMsg'
Learn More

Ergosterol, TRC

Product Code. 17967724
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17967724 1 g 1g
17948984 100 mg 100mg
17929644 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17967724 Supplier Toronto Research Chemicals Supplier No. TRCE5992401G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-87-4
Molecular Formula C28 H44 O
Molecular Weight (g/mol) 396.65
Synonym Ergosta-5,7,22-trien-3-ol, (3β,22E)-, Ergosterol (8CI), (3β,22E)-Ergosta-5,7,22-trien-3-ol, (22E,24R)-Ergosta-5,7,22-trien-3β-ol, (24R)-Ergosta-5,7,22-trien-3β-ol, 24-Methylcholesta-5,7,22-trien-3β-ol, 24R-Methylcholesta-5,7,22E-trien-3β-ol, 24α-Methyl-22E-dehydrocholesterol, 3β-Hydroxyergosta-5,7,22-triene, Ergosterin, Provitamin D, Provitamin D2
IUPAC Name (3S,9S,10R,13R,14R,17R)-17-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H44O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-10,18-20,22,24-26,29H,11-17H2,1-6H3/b8-7+/t19-,20+,22-,24+,25-,26-,27-,28+/m0/s1
Formula Weight 396.3392
Percent Purity >95%
Chemical Name or Material Ergosterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.