missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome™ Black T, Indicator, Reag. Ph. Eur. for metal titration, Solstice

Product Code. 15633850 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15633850 25 g Glass Bottle 25g
15613850 100 g Glass Bottle 100g
15623850 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15633850 Supplier Honeywell-Fluka Supplier No. 3275125G
Quantity Price
1 40.05 € / 25g
10 37.07 € / 25g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator, Reag. Ph. Eur. for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
Synonym Mordant Black 11
PubChem CID 87355429
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

CAS Max % 100.0000%
Packaging Glass Bottle
Suitability Complies for indicator
Linear Formula C20H12N3NaO7S
UN Number NONH for all modes of transport
Beilstein 4121162
Quantity 25 g
IUPAC Name (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
Formula Weight 461.38
Grade Ph. Eur.
Chemical Name or Material Eriochrome™ Black T, Indicator, Reag. Ph. Eur. for metal titration
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.