missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Black T, Pure, C.I.14645, Metal Indicator, Fisher Chemical™

Product Code. 10793981 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10793981 25 g 25g
10467060 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10793981 Supplier Fisher Chemical Supplier No. E/0100/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eriochrome Black T, Pure, C.I.14645, Metal Indicator

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
PubChem CID 87355429
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

IUPAC Name (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
Formula Weight 461.38g/mol
Packaging Glass Jar
Color Black
pH 3.7
Quantity 25 g
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • Eriochrome Black T
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.