missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eriochrome™ Black T, pure, indicator grade

Product Code. 10460121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10460121 25 g Glass bottle 25g
10793421 100 g Glass bottle 100g
10073110 500 g Glass bottle 500g
10256570 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10460121 Supplier Thermo Scientific Chemicals Supplier No. 228360250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1787-61-7 | C20H12N3NaO7S | 461.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
Synonym C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11
PubChem CID 87355429
IUPAC Name (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Beilstein 16, III, 320
Merck Index 15, 3725
Solubility Information Solubility in water: slightly soluble.
Infrared Spectrum Authentic
Formula Weight 461.38
Grade Pure
Packaging Glass bottle
Suitability Passes (for complexometric determination of metals)
UV Spectrometry Complies
Color Black to Black/Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Eriochrome™ Black T
Show More Show Less
compliance-icons
Product Identifier
  • Eriochrome Black T
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.