missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome™ Blue Black B, Honeywell™ Fluka™

Product Code. 15695560 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 15695560

Brand: Honeywell-Fluka 4554025G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 3564-14-5
Molecular Formula C20H13N2NaO5S
MDL Number MFCD00003934
UN Number NONH for all modes of transport
InChI Key YRFCCVCSEVISHI-GXTSIBQPSA-M
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O
IUPAC Name sodium;4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
Molecular Weight (g/mol) 416.38
PubChem CID 23722702
Formula Weight 416.38g/mol
Packaging Glass bottle
Color Black
pH 9.5 (at 10 g/L 20°C)
Quantity 25 g
Physical Form Powder
Chemical Name or Material Eriochrome™ Blue Black B
Show More Show Less
compliance-icons
Product Identifier
  • Eriochrome Blue Black B
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium 3-hydroxy-4-[(1-hydroxy-2-naphthyl)azo]naphthalene-1-sulphonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.