missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eriochrome∣r Blue Black B

Product Code. p-7042692
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15485639

Brand: Thermo Scientific Chemicals J63396.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3564-14-5 | C20H13N2NaO5S | 416.38 g/mol

Eriochrome is to do titration indicator. Dubbed in aqueous solution or alcohol-soluble when used for the determination of calcium, magnesium, manganese, zinc and other metal ions, measured water hardness.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eriochrome is to do titration indicator. Dubbed in aqueous solution or alcohol-soluble when used for the determination of calcium, magnesium, manganese, zinc and other metal ions, measured water hardness.

Solubility
Soluble in water(20 g/L (20 C).

Notes
Eriochrome is a registered trademark of Ciba Specialty Chemicals Corp.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3564-14-5
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.38
MDL Number MFCD00003934
InChI Key YRFCCVCSEVISHI-GXTSIBQPSA-M
Synonym C.I. 14640; Mordant Black 3
PubChem CID 23722702
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O

Specifications

Solubility Information Soluble in water(20g/L (20 C).
IUPAC Name sodium;4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
Formula Weight 416.39
Color Black to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Eriochrome™ Blue Black B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.