missing translation for 'onlineSavingsMsg'
Learn More

Erioglaucine sodium salt

Product Code. 15434299
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434299 25 g 25g
15424299 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434299 Supplier Thermo Scientific Alfa Aesar Supplier No. J61780.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3844-45-9 | C37H34N2Na2O9S3 | 792.844 g/mol

Erioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.

Solubility
Soluble in water (30 mg/ml), ethanol.

Notes
Dye content: approx 60%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3844-45-9
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012141
InChI Key SGHZXLIDFTYFHQ-UHFFFAOYSA-L
Synonym C.I. 42090; Acid Blue 9
PubChem CID 19700
ChEBI CHEBI:82411
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 283°C
Merck Index 14,1373
Solubility Information Soluble in water (30mg/ml),ethanol.
Formula Weight 792.86
Color Purple
Quantity 25 g
Physical Form Powder
Chemical Name or Material Erioglaucine sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.