Learn More
Heinz Herenz Erlenmeyer Caps
Product Code. p-200111932
Click to view available options
Color:
Blue
Red
Silver
Unit Size:
Pack of 10
Product Code. 30158407
Brand: Heinz Herenz 1080926
Only null left
Add to basket
Add to basket
Erlenmeyer cap without handles.
  • Erlenmeyer caps
  • Aluminium
  • 38 mm
  • Without handle
Specifications
Color Red
Product Type Cap without Handle
Autoclavable Autoclavable
Disposable Reusable
Sterility Non-sterile
Closure Type Straight Rimless Neck
Diameter (Metric) Outer 38 mm
Closure Size 38 mm