missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Erlenmeyer flask

Titrate chemical samples without spillage. Hirschmann™ Erlenmeyer flask is ideal for performing titration and preparing microbial cultures.

Brand:  Hirschmann™ 7630180

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10054424

  • 39.90 € / Pack of 6

Estimated Shipment: 28-11-2023
to see stock.
Add to basket

Description

Description

  • Wide neck
  • Standard ground PP stoppers
  • Transparent
  • With PP screw cap
Specifications

Specifications

100 mL
20 mL
NS 34/35
Yes
Polypropylene
Wide
40%
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers