missing translation for 'onlineSavingsMsg'
Learn More

ERM (LRRKtide) Substrate

Product Code. 10481695
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10481695 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10481695 Supplier Invitrogen Supplier No. PV5093
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ERM (LRRKtide) Substrate

A peptide substrate based upon ezrin/radixin/moesin (ERM) that is phosphorylated by LRRK2 and LRRK2 mutants. For use with the Adapta™ Universal Kinase Assay (PV5099).
TRUSTED_SUSTAINABILITY

Specifications

Amino Acid Sequence RLGRDKYKTLRQIRQ
Assay Entry Biochemical ADP accumulation
Content And Storage 2 mM ERM (LRRKtide) peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA. Store at -20°C.
Detection Method Fluorescence
Quantity 1 mL
Substrate peptide based upon ezrin/radixin/moesin (ERM)
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 120892
Molecular Weight (g/mol) 1931.2 Da
Shipping Condition Dry Ice
Target Entry LRRK2 G2019S, LRRK2
Validated Kinases LRRK2
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.