missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ERN2, GST Tag Recombinant Protein
This ERN2 Recombinant Human Protein (499-end) was expressed in insect cells.
Brand: Invitrogen™ A32890
Additional Details : Weight : 0.01000kg
Description
- This ERN2 recombinant human protein (499–end) was expressed in insect cells
- ERN2 serine/threonine kinase, or endoplasmic reticulum to nucleus signaling 2 protein, is a human homologue of the yeast Ire1 gene product
- It plays a role in multiple facets of the ER stress response in mammalian cells
- ERN2 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
NP_150296 | |
79.6 kDa | |
IRE Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
100 μg | |
aa 499 - end | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Recombinant, Active, Catalytic |