missing translation for 'onlineSavingsMsg'
Learn More

Erucic Acid Methyl Ester, TRC

Product Code. 30135383
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135383 25 mL 25mL
30133803 5 mL 5mL
30005828 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135383 Supplier Toronto Research Chemicals Supplier No. TRCE64991525ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-34-9
Molecular Formula C23 H44 O2
Molecular Weight (g/mol) 352.59
Synonym Methyl Erucate, 13-Docosenoic acid, methyl ester, (13Z)-, 13-Docosenoic acid, methyl ester, (Z)- (8CI), Brassidic acid, methyl ester (6CI), (Z)-Erucic acid methyl ester, Agnique ME 22U, Erucic acid methyl ester, Methyl 13(Z)-docosenoate, Methyl cis-13-docosenoate, Methyl erucate
IUPAC Name methyl (Z)-docos-13-enoate
SMILES CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OC

Specifications

Quantity 25 mL
InChI Formula InChI=1S/C23H44O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h10-11H,3-9,12-22H2,1-2H3/b11-10-
Formula Weight 352.3341
Chemical Name or Material Erucic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.