missing translation for 'onlineSavingsMsg'
Learn More

Erucic acid, tech. 90%

Product Code. 11342656
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342656 250 g 250g
11332656 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342656 Supplier Thermo Scientific Alfa Aesar Supplier No. L08685.30
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-86-7 | C22H42O2 | 338.576 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.576
MDL Number MFCD00063188
InChI Key DPUOLQHDNGRHBS-KTKRTIGZSA-N
Synonym erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
PubChem CID 5281116
ChEBI CHEBI:28792
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)O

Specifications

Melting Point 29°C to 33°C
Boiling Point 265°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
Beilstein 1728049
Merck Index 14,3674
Formula Weight 338.58
Percent Purity 90%
Grade Technical
Chemical Name or Material Erucic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.