missing translation for 'onlineSavingsMsg'
Learn More

Erucic acid, tech. 90%

Product Code. p-7044908
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11332656

Brand: Thermo Scientific Alfa Aesar L08685.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-86-7 | C22H42O2 | 338.576 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.576
MDL Number MFCD00063188
InChI Key DPUOLQHDNGRHBS-KTKRTIGZSA-N
Synonym erucic acid, cis-13-docosenoic acid, z-docos-13-enoic acid, 13-docosenoic acid, z, cis-erucic acid, 13-cis-docosenoic acid, 13-docosenoic acid, z-13-docosenoic acid, hystrene 2290
PubChem CID 5281116
ChEBI CHEBI:28792
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)O

Specifications

Melting Point 29°C to 33°C
Boiling Point 265°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 1728049
Merck Index 14,3674
Formula Weight 338.58
Percent Purity 90%
Grade Technical
Chemical Name or Material Erucic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.