missing translation for 'onlineSavingsMsg'
Learn More

erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide, 95%, Thermo Scientific Chemicals

Product Code. 15489575
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489575 1 g 1g
15499575 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489575 Supplier Thermo Scientific Alfa Aesar Supplier No. H52099.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 388071-27-0
Molecular Formula C15H19F2NO3
Molecular Weight (g/mol) 299.32
MDL Number MFCD08061630
InChI Key NKGKCDXMOMAORK-UHFFFAOYNA-N
Synonym erythro-n-boc-l-3,5-difluorophenylalanine epoxide, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethyl carbamate, tert-butyl n-1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate, erythro-n-boc-3,5-difluoro-l-phenylalanine epoxide, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiranyl ethylcarbamate, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate
PubChem CID 9922319
IUPAC Name tert-butyl N-[(1S)-2-(3,5-difluorophenyl)-1-[(2S)-oxiran-2-yl]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC(F)=CC(F)=C1)C1CO1

Specifications

Quantity 1 g
Formula Weight 299.31
Percent Purity 95%
Chemical Name or Material erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.