missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Erythroid Progenitor Reprogramming Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- The Erythroid Progenitor Reprogramming Kit contains an integrated set of tools and reagents to enrich, expand and reprogram erythroid progenitor cells from peripheral blood
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells, Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Reprogramming |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction