missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin A 6,9-Imino Ether, TRC

Product Code. 30011821
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011821 100 mg 100mg
30011343 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011821 Supplier Toronto Research Chemicals Supplier No. TRCE650015100MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99290-97-8
Molecular Formula C37 H66 N2 O12
Molecular Weight (g/mol) 730.93
Synonym 7,16-Dioxa-2-azabicyclo[11.2.1]hexadec-1-en-8-one, 10-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-6-ethyl-4,5-dihydroxy-3,5,9,11,13,15-hexamethyl-12-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, (1Z,3R,4R,5S,6R,9R,10S,11S,12R,13R,15R)-, 7,16-Dioxa-2-azabicyclo[11.2.1]hexadec-1-en-8-one, 10-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-6-ethyl-4,5-dihydroxy-3,5,9,11,13,15-hexamethyl-12-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, [3R-(3R*,4R*,5S*,6R*,9R*,10S*,11S*,12R*,13R*,15R*)]-, (1Z,3R,4R,5S,6R,9R,10S,11S,12R,13R,15R)-10-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-6-ethyl-4,5-dihydroxy-3,5,9,11,13,15-hexamethyl-12-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-7,16-dioxa-2-azabicyclo[11.2.1]hexadec-1-en-8-one
IUPAC Name (1Z,3R,4R,5S,6R,9R,10S,11S,12R,13R,15R)-12-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyl-tetrahydropyran-2-yl]oxy-6-ethyl-4,5-dihydroxy-10-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyl-tetrahydropyran-2-yl]oxy-3,5,9,11,13,15-hexamethyl-7,16-dioxa-2-azabicyclo[11.2.1]hexadec-1-en-8-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@]4(C)C[C@@H](C)\C(=N\[C@H](C)[C@@H](O)[C@]1(C)O)\O4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C37H66N2O12/c1-14-25-37(10,44)29(41)22(6)38-32-18(2)16-36(9,51-32)31(50-34-27(40)24(39(11)12)15-19(3)46-34)20(4)28(21(5)33(43)48-25)49-26-17-35(8,45-13)30(42)23(7)47-26/h18-31,34,40-42,44H,14-17H2,1-13H3/b38-32-/t18-,19-,20+,21-,22-,23+,24+,25-,26+,27-,28+,29-,30+,31-,34+,35-,36-,37-/m1/s1
Formula Weight 730.4616
Percent Purity >95%
Chemical Name or Material Erythromycin A 6,9-Imino Ether
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.