missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine-d10-N-FMOC, CDN

Product Code. 30314533
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30314533

Brand: CDN CDND54920.1G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-2,3,3,4,5,5,5,5',5',5'-D10
CAS 1190594-22-9
Molecular Formula C21 D10 H13 N O4
Quantity 0.1 g
Synonym L-Leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (ACI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10 (ACI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-2,3,3,4,5,5,5,5',5',5'-D10, L-Leucine-d10-N-FMOC, (2 S)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-(methyl-d3)pentanoic-2,3,3,4,5,5,5-d7 acid, N-FMOC-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-FMOC-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-FMOC-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10
InChI Formula InChI=1 S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19 H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m0/s1/i1D3,2D3,11D2,13 D,19 D
SMILES [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[C@]([2 H])(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC Name (2 S)-2,3,3,4,5,5,5-heptadeuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-4-(trideuteriomethyl)pentanoic acid
Molecular Weight (g/mol) 363.4732
Formula Weight 363.2255 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.