missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Erythrosin B, pure, certified

Product Code. 10299450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10299450 25 g Glass bottle 25g
10289033 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10299450 Supplier Thermo Scientific Chemicals Supplier No. 409450250
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16423-68-0 | C20H6I4Na2O5 | 879.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16423-68-0
Molecular Formula C20H6I4Na2O5
Molecular Weight (g/mol) 879.86
MDL Number MFCD00144257
InChI Key IINNWAYUJNWZRM-UHFFFAOYSA-L
Synonym Acid Red 51, C.I. 45430
PubChem CID 12961638
SMILES [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12

Specifications

Absorption Ratio 1.15 to 1.55
Dye Content 80% min.
Infrared Spectrum Authentic
Lambda Max 524 to 527nm
Packaging Glass bottle
Beilstein 19, II, 258
Merck Index 15, 3749
Color Brown to Red
Quantity 25 g
Physical Form Fine Powder
IUPAC Name disodium;2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
Formula Weight 879.85
Grade Pure
Chemical Name or Material Erythrosin B, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Erythrosin B
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.