missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B

Product Code. 11442247
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442247 100 g 100g
11432247 25 g 25g
11452247 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442247 Supplier Thermo Scientific Alfa Aesar Supplier No. A14180.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Erythrosin B, C40H14I8Na2O10, CAS Number-16423-68-0, C.I. 45430; Tetraiodofluorescein sodium salt, 500g, 240-046-0, 879.87, H302, GHS H Statement H302 Harmful if swallowed., P264b-P270-P301+P312-P330-P501c, DKRYETBOLGDIOK-UHFFFAOYSA-L | CAS: 16423-68-0 | C40H14I8Na2O10 | 1715.757 g/mol

Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

Solubility
Soluble in water, glycerin and methanol.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16423-68-0
MDL Number MFCD00144257
Synonym C.I. 45430; Tetraiodofluorescein sodium salt
PubChem CID 131676154

Specifications

Sensitivity Hygroscopic; Light sensitive
Molecular Formula C40H14I8Na2O10
Solubility Information Soluble in water,glycerin and methanol.
Quantity 100 g
InChI Key DKRYETBOLGDIOK-UHFFFAOYSA-L
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate
Molecular Weight (g/mol) 1715.757
Formula Weight 879.87
Chemical Name or Material Erythrosin B

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.