missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ ES 7X™-PF Cleaning Solution

Product Code. 10019520 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (English):
1 gal.
1.32
4 x 1 gal.
5 gal.
55 gal.
Unit Size:
10 L
18.92 L
208.19 L
Each
Pack of 4
Pack of 5
Pack of 55
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (English) Unit Size
10019520 1.32 10 L
11409585 4 x 1 gal. Pack of 4
11419585 5 gal. Pack of 5
17104185 5 gal. 18.92 L
11499575 1 gal. Each
11429585 55 gal. Pack of 55
17114185 55 gal. 208.19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10019520 Supplier MP Biomedicals™ Supplier No. 097667121
  • ONLINE EXCLUSIVE PRICE
    217.00 €
    205.80 € / 10 L
    Save 11.20 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

  • Safe and effective
  • Soluble in water at any concentration
  • Drains from glassware in seconds
  • Contains powerful sequestering reagents
  • Ideal for cell cultureware and general labware
  • Cleans both glass and plastic labware

Specifications

Product Type Phosphate-free Cleaning Solution
Quantity 2 Pk.
Volume (English) 1.32
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • ES-7X Cleaning Solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Dioctyl Sulfosuccinate Sodium Salt, 2-(2-butoxyethoxy)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.