missing translation for 'onlineSavingsMsg'
Learn More

Melford Esculin

Product Code. p-200087177
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 30018253

Brand: Melford E3800025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • CAS Number: 531-75-9
  • Molecular Formula: C15H16O9 xH2O
  • Molecualr Weight: 340.28 g/mol

Specifications

CAS 531-75-9
Quantity 25 g
Molecular Formula C15H16O9 - xH2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.