missing translation for 'onlineSavingsMsg'
Learn More

Esculin sesquihydrate, 97%

Product Code. 11494713
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494713 25 g 25g
11484713 5 g 5g
11404723 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494713 Supplier Thermo Scientific Alfa Aesar Supplier No. A11624.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66778-17-4 | C30H38O21 | 734.613 g/mol

Esculin sesquihydrate is used to inhibits chemically induced carcinogenesis in mouse skin and kidney, used in a microbiology laboratory to aid in the identification of bacterial species, a naturally occurring antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Esculin sesquihydrate is used to inhibits chemically induced carcinogenesis in mouse skin and kidney, used in a microbiology laboratory to aid in the identification of bacterial species, a naturally occurring antioxidant.

Solubility
Slightly soluble in water.

Notes
Air Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66778-17-4
Molecular Formula C30H38O21
Molecular Weight (g/mol) 734.613
MDL Number MFCD00149492
InChI Key LGMHQDJZWHRZPO-HPPUHPNBSA-N
Synonym esculin sesquihydrate
PubChem CID 131675424
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
SMILES C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.O.O.O

Specifications

Melting Point 203°C to 205°C
Assay Percent Range 97%
Quantity 25 g
Beilstein 95387
Merck Index 14,3698
Solubility Information Slightly soluble in water.
Formula Weight 367.31 (340.28 Anhydrous)
Percent Purity 97%
Chemical Name or Material Esculin Sesquihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.