Learn More
Notrax ESD Common Grounding Cord
Product Code. 17711918
Change view
Click to view available options
Connector Types:
Female Connector
Ring Connector
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
17711918 Ring Connector Each
17721918 Female Connector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17711918 Supplier Notrax Supplier No. 052SCG0100
  • ONLINE EXCLUSIVE PRICE
    24.15 €
    20.90 € / Pack of 1
    Save 3.25 €
    13% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment.
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
  • 45 m cable connects wrist bands or bench mat to ground
  • Assures that all parts are at the same ground potential
  • Easy to install, simply snap it to a mat, plug in the wrist strap coil cord and connect terminal to ground
  • Resistor 1 Meg mat to ground
  • Available with a ring terminal or 10 mm female socket
Specifications
Color Black
Product Type Grounding Cord
Material Wire
Connector Types Ring Connector