Learn More
Notrax ESD Non-Abrasive Mat Cleaner
Product Code. 17752488
Change view
Click to view available options
Product Type:
Mat Cleaner
Unit Size:
Each
Product Code. Product Type Unit Size
17752488 Mat Cleaner Each
1 options
Product Code. 17752488 Supplier Notrax Supplier No. 057SMC0100
  • ONLINE EXCLUSIVE PRICE
    30.45 €
    26.62 € / Pack of 1
    Save 3.83 €
    12% Off
View all promotions
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Mat cleaner is recommended for static safe table and floor mats. Its non-abrasive formula leaves no film and is designed to enhance the static dissipative properties of ESD safe surfaces.
  • Mat cleaner is recommended for static safe table and floor mats
  • Its non abrasive formula leaves no film and is designed to enhance the static dissipative properties of
  • ESD safe surfaces
  • Effectively removes flux residues, oil, finger marks and dirt
  • Convenient 1000 mL bottle trigger spray
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
Specifications
Product Type Mat Cleaner
Material Wire
For Use With (Application) Remove Flux Residues, Oil, Finger Marks and Dirt