Learn More
uvex ESS Hypervision CA FBL Antifog
Product Code. 31751171 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751171 Polycarbonate Each
1 options
Product Code. 31751171 Supplier uvex Supplier No. 9321655
  • ONLINE EXCLUSIVE PRICE
    11.44 €
    10.96 € / Pack of 1
    Save 0.48 €
    4% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to restore visibility and extend the usable life of safety spectacles through sustainable replacement.
  • Act sustainably by using our replacement lenses to extend the life of your safety spectacles. Scratched lens? No problem-simply replace the lens and continue wearing the same safety spectacles.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Replacement Lens
Disposable Reusable
Lens Material Polycarbonate