missing translation for 'onlineSavingsMsg'
Learn More

Estradiol 17-Isovalerate, TRC

Product Code. 30374521
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374521 25 mg
30371026 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374521 Supplier Toronto Research Chemicals Supplier No. TRCE88806025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869627-83-8
Molecular Formula C23H32O3
Molecular Weight (g/mol) 356.51
Synonym (17beta)-Estra-1,3,5(10)-triene-3,17-diol 17-(3-Methylbutanoate)
SMILES CC(C)CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H32O3/c1-14(2)12-22(25)26-21-9-8-20-19-6-4-15-13-16(24)5-7-17(15)18(19)10-11-23(20,21)3/h5,7,13-14,18-21,24H,4,6,8-12H2,1-3H3/t18-,19-,20+,21+,23+/m1/s1
Formula Weight 356.2351
Chemical Name or Material Estradiol 17-Isovalerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.