missing translation for 'onlineSavingsMsg'
Learn More

Estradiol 3-Sulfate 17Beta-Glucuronide Dipotassium Salt, TRC

Product Code. 30384442
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384442 2.5 mg 2.5mg
30369487 1 mg 1mg
30384823 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384442 Supplier Toronto Research Chemicals Supplier No. TRCE8888302.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10392-35-5
Molecular Formula C24 H30 O11 S . 2 K
Molecular Weight (g/mol) 604.75
Synonym Estradiol 3-Sulfate 17-B-Glucuronide Dipotassium Salt
IUPAC Name dipotassium;(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[[(8R,9S,13S,14S,17S)-13-methyl-3-sulfonatooxy-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl]oxy]oxane-2-carboxylate
SMILES [K+].[K+].C[C@]12CC[C@H]3[C@@H](CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CC[C@@H]2O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C24H32O11S.2K/c1-24-9-8-14-13-5-3-12(35-36(30,31)32)10-11(13)2-4-15(14)16(24)6-7-17(24)33-23-20(27)18(25)19(26)21(34-23)22(28)29;;/h3,5,10,14-21,23,25-27H,2,4,6-9H2,1H3,(H,28,29)(H,30,31,32);;/q;2*+1/p-2/t14-,15-,16+,17+,18+,19+,20-,21+,23-,24+;;/m1../s1
Formula Weight 604.0783
Chemical Name or Material Estradiol 3-sulfate 17beta-Glucuronide dipotassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.