missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals β-Estradiol, 98%

Product Code. 11916891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11916891 5 g Glass bottle 5g
11906891 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916891 Supplier Thermo Scientific Chemicals Supplier No. 436320050
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

β-Estradiol, 98%, CAS # 50-28-2, is an estrogen steroid sex hormone that regulates several biological functions including gene transcription and protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• β-Estradiol is a naturally occurring endogenous hormone in females

• It is an important steroid sex hormone involved in the regulation of several biological functions including gene transcription and protein synthesis

• This hormone is produced by the ovaries and can bind to and activate specific nuclear receptors

Applications

• β-Estradiol acts as an estrogenic hormone receptor agonist and activator of PI 3-kinase

• It shows inhibitory activity against aldehyde oxidase

• β-Estradiol presents mild anabolic and metabolic activities and increases blood coagulability in vivo experiments

• It plays a role in immune and inflammatory processes

• β-Estradiol triggers an increase in hepatic synthesis of some proteins, including sex hormone-binding globulin (SHBG), and thyroid-binding globulin (TBG), and suppresses the formation of follicle-stimulating hormone (FSH)

• This compound has been used for the in vitro maturation of bovine cumulus-oocyte complexes (COCs)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-28-2
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.38
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
PubChem CID 5757
ChEBI CHEBI:16469
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 176.0°C to 180.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,3758
Specific Rotation +76° to +83° (20°C, 589nm) (c=1, dioxane)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, hexane, ethyl acetate
Formula Weight 272.38
Percent Purity 98%
Physical Form Powder
Chemical Name or Material β-Estradiol
Show More Show Less
compliance-icons
Product Identifier
  • beta-Estradiol
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H362-May cause harm to breast-fed children.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P263-Avoid contact during pregnancy/while nursing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.