missing translation for 'onlineSavingsMsg'
Learn More

β-Estradiol, MP Biomedicals™

Product Code. 11404942 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404942 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404942 Supplier MP Biomedicals Supplier No. 02194565.2
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

β-Estradiol is a potent mammalian estrogenic hormone produced by the ovary. Triggers the production of gonadotropins, leading to ovulation.

Estrogens direct the development of the female phenotype in embryogenesis and during puberty by regulating gene transcription and, thus, protein synthesis. It also induces the production of gonadotropins which, in turn, induce ovulation.

  • The major estrogen secreted by the premenopausal ovary.
  • Exposure to estradiol increases breast cancer incidence and proliferation.

Specifications

CAS 50-28-2
Melting Point 173°C to 179°C
Molecular Formula C18H24O2
Quantity 250 mg
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
Solubility Information Almost insoluble in water (50mg/mL; clear, colorless solution). Freely soluble in alcohol; soluble in acetone, dioxane, other organic solvents; solutions of fixed alkali hydroxides; sparingly soluble in vegetable oils; soluble in methanol:CHCl3 (1:1).
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
Optical Rotation +76.8 (Concentration = 1, Solvent = Dioxane) (Lit.)
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 272.388
PubChem CID 5757
ChEBI CHEBI:16469
Formula Weight 272.39g/mol
Grade Cell Culture Reagent
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.