missing translation for 'onlineSavingsMsg'
Learn More

β-Estradiol, MP Biomedicals

Product Code. 11488020 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488020 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11488020 Supplier MP Biomedicals Supplier No. 02101656.2
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • β-Estradiol is used to study cell differentiation and transformations (tumorigenicity)
  • The pharmacology and HPLC determination of β-Estradiol in plasma and urine have been reported

Specifications

CAS 50-28-2
Melting Point 173°C to 179°C
Color White
Molecular Formula C18H24O2
Linear Formula C18H24O2
MDL Number MFCD00003693
Quantity 250 mg
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
Solubility Information Almost insoluble in water(50mg/mL-clear, colorless solution). Freely soluble in alcohol; soluble in acetone, dioxane, other organic solvents; solutions of fixed alkali hydroxides; sparingly soluble in vegetable oils; soluble in methanol:CHCl3 (1:1)
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 272.388
PubChem CID 5757
ChEBI CHEBI:16469
Formula Weight 272.39
Physical Form Powder
Chemical Name or Material β-ESTRADIOL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.