missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Estradiol Parameter Assay Kit

Product Code. 18752144
Change view
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752144 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18752144 Supplier R&D Systems™ Supplier No. KGE014
  • ONLINE EXCLUSIVE PRICE
    587.26 €
    523.95 € / 1 sets
    Save 63.31 €
    10% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Estradiol Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Estradiol Parameter Assay Kit quantifies multi-species Estradiol in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 12.30 - 3000pg/mL
Assay Sensitivity 12.1pg/mL
Conjugate Biotin
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Estradiol levels in various samples. This assay cross-reacts 0.07% with 17 alpha-ethynylestradiol, 0.03% with 17 beta-estradiol 3-Benzoate, 0.86% with Estriol, 0.26% with Estrone and 0.06% with Progesterone.
For Use With (Application) ELISA
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.