missing translation for 'onlineSavingsMsg'
Learn More

Estriol, TRC

Product Code. 18062120
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18062120 100 mg 100mg
18061730 1 g 1g
18011040 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18062120 Supplier Toronto Research Chemicals Supplier No. TRCE888960100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-27-1
Molecular Formula C18 H24 O3
Molecular Weight (g/mol) 288.38
Synonym Estriol, (16α,17β)-Estra-1,3,5(10)-triene-3,16,17-triol, 13β-Methyl-1,3,5(10)-gonatriene-3,16α,17β-triol, 16α,17β-Estriol, 16α-Estriol, 16α-Hydroxyestradiol, 3,16α,17β-Estriol, 3,16α,17β-Trihydroxyestra-1,3,5(10)-triene, Aacifemine, Colpogyn, Destriol, Estra-1,3,5(10)-triene-3,16α,17β-triol, Estratriol, Follicular hormone hydrate, Gynasan, Holin V, Hormomed, Incurin, Klimax E, Klimoral, NSC 12169, OE3, Oekolp, Oestriol, Ortho-Gynest, Ovesterin, Ovestin, Ovo-Vinces, Theelol, Tridestrin, Trihydroxyestrin, Triovex
IUPAC Name (8R,9S,13S,14S,16R,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1
Formula Weight 288.1725
Percent Purity >95%
Chemical Name or Material Estriol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.