missing translation for 'onlineSavingsMsg'
Learn More

Estriol Trisulfate Trisodium Salt, TRC

Product Code. 30004262
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004262 25 mg 25mg
30011989 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004262 Supplier Toronto Research Chemicals Supplier No. TRCE88897025MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100940-55-4
Molecular Formula C18H21Na3O12S3
Molecular Weight (g/mol) 594.52
SMILES [Na+].[Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1C[C@@H](OS(=O)(=O)[O-])[C@@H]2OS(=O)(=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H24O12S3.3Na/c1-18-7-6-13-12-5-3-11(28-31(19,20)21)8-10(12)2-4-14(13)15(18)9-16(29-32(22,23)24)17(18)30-33(25,26)27;;;/h3,5,8,13-17H,2,4,6-7,9H2,1H3,(H,19,20,21)(H,22,23,24)(H,25,26,27);;;/q;3*+1/p-3/t13-,14-,15+,16-,17+,18+;;;/m1.../s1
Formula Weight 593.9888
Chemical Name or Material Estriol Trisulfate Trisodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.