missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estrone-2,4,16,16-d4, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Estrone D4 |
| CAS | 53866-34-5 |
| Molecular Formula | C18 D4 H18 O2 |
| Quantity | 0.01 g |
| Synonym | Estrone-D4, Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-hydroxy-, 3-Hydroxyestra-1,3,5(10)-trien-17-one-2,4,16,16-d4, Estrone-2,4,16,16-d4, Estrone-(2,4,16,16-D4), Estrone-2,4,16,16-D4 |
| InChI Formula | InChI=1 S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19 H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1/i3D,7D2,10 D |
| SMILES | [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])C4=O)[C@@H]3 CCc2c([2 H])c1O |
| IUPAC Name | (8 R,9 S,13 S,14 S)-2,4,16,16-tetradeuterio-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 274.391 |
| Formula Weight | 274.187 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction