missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Estrone-3-Glucuronide (E1G) Competitive ELISA Kit

Product Code. 15805853
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15805853 96 Tests Pack of 96
17865117 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15805853 Supplier Invitrogen™ Supplier No. EIA17E3
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ELISA

The Estrone-3 Glucuronide (E1G) ELISA quantitates estrone-3 glucuronide in extracted serum,plasma,urine,dried fecal extracts or cell culture medium.

Principle of the method

The Estrone-3-Sulfate (E1S) Competitive ELISA research-use-only kit uses a specifically generated antibody to measure Estrone-3-Sulfate (E1S) in a variety of matrices,including serum,plasma,urine,and fecal samples. The kit will quantitatively measure E1S present in diluted buffer samples and tissue culture media samples. An E1S standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An E1S-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to E1S to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound E1S-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Estrone-3-glucuronide (E1G) is the principle secreted form of circulating estradiol in mammals. Estrone-3-glucuronide (E1G) is identical across all species.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 7.38 pg/mL
Description Estrone-3-Glucuronide (E1G) Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA
Protein Family Steroid hormone
Sample Type Fecal Extract, Serum, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 5.6%
Intraassay CV 3.8
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 6.25 μL
Storage Requirements -20°C
Target Estrone-3-Glucuronide
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.