Learn More
Cole-Parmer™ Etatron DLXB pH/ORP Pump Control System
Product Code. 15813147 Shop All Cole-Parmer Products
Click to view available options
Flow Rate:
0.26 GPH
Unit Size:
Each
Product Code. 15813147
Brand: Cole-Parmer™ 5602287
Only null left
Add to basket
Add to basket
Control the pH or ORP levels of your process with the integrated Etatron Pump Control System. Saves you time, money, and space.
  • Integrated controller and metering pump simplify system setup
    • Control systems are able to accept pH or ORP electrodes for added convenience
    • On/off or proportional control modes allow for robust process control
    • Deadband is programmable for greater flexibility
    • LCD is easy to read
Specifications
Includes 6 ft. (1.8 m) Clear PVC and PE Tubing (3⁄16 x 1⁄4 in.), Weighted Filter, and 6 ft. (1.8 m) U.S. Standard Power Cord
Flow Rate 0.26 GPH
Frequency 50/60 Hz
Maximum Pressure 217.5 psi
Voltage 230 V