missing translation for 'onlineSavingsMsg'
Learn More

Ethacridine Lactate Monohydrate, TRC

Product Code. 30382730
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382730 5 g
30382590 10 g
30369375 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382730 Supplier Toronto Research Chemicals Supplier No. TRCE6760235G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6402-23-9
Molecular Formula C15 H15 N3 O . C3 H6 O3 . H2 O
Molecular Weight (g/mol) 361.39
Synonym Ethacridine Lactate Monohydrate, 7-Ethoxyacridine-3,9-diamine (2RS)-2-hydroxypropanoate monohydrate, 7-Ethoxy-3,9-acridinediamine lactate monohydrate, 6,9-Diamino-2-ethoxyacridine lactate monohydrate, 2,5-Diamino-7-ethoxyacridine lactate monohydrate, 2-Ethoxy-6,9-diaminoacridine lactate monohydrate, Etakridin lactate monohydrate, Acrinol, Rimaon, Rivanol, Vucine, Acrolactine, Ethodin, Metifex
IUPAC Name 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate
SMILES O.CCOc1ccc2nc3cc(N)ccc3c(N)c2c1.CC(O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H15N3O.C3H6O3.H2O/c1-2-19-10-4-6-13-12(8-10)15(17)11-5-3-9(16)7-14(11)18-13;1-2(4)3(5)6;/h3-8H,2,16H2,1H3,(H2,17,18);2,4H,1H3,(H,5,6);1H2
Formula Weight 361.1638
Chemical Name or Material Ethacridine lactate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.